Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-chlorobenzoic acid, 98+%

$33.24 - $352.50

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-chlorobenzoic acid
CAS 59748-90-2
Molecular Formula C7H4BrClO2
Formula Weight 235.47
MDL Number MFCD00040903
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1152703

 
alfa aesar™
A1152703
1g Each for $33.24

AAA1152706

 
alfa aesar™
A1152706
5g Each for $107.12

AAA1152714

 
alfa aesar™
A1152714
25g Each for $352.50
Description & Specifications

Specifications

Formula Weight 235.47
Chemical Name or Material 4-Bromo-2-chlorobenzoic acid
Assay ≥98%
Molecular Formula C7H4BrClO2
MDL Number MFCD00040903
CAS 59748-90-2
Melting Point 172° to 174°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.