Learn More
4-Bromo-2-chlorobenzoic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1152706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Bromo-2-chlorobenzoic acid | |
172°C to 174°C | |
MFCD00040903 | |
benzoic acid, 4-bromo-2-chloro, 4-bromo-2-chlorobenzoicacid, 2-chloro-4-bromobenzoic acid, 4-bromo-2-chloro-benzoic acid, rarechem al bo 2371, attercop-chm at111793, zlchem 410, pubchem3586, acmc-1b03i, ksc274c2d | |
C1=CC(=C(C=C1Br)Cl)C(=O)O | |
235.461 | |
235.47 |
59748-90-2 | |
C7H4BrClO2 | |
5 g | |
JAVZWSOFJKYSDY-UHFFFAOYSA-N | |
4-bromo-2-chlorobenzoic acid | |
108829 | |
≥98% |
Chemical Identifiers
59748-90-2 | |
235.461 | |
JAVZWSOFJKYSDY-UHFFFAOYSA-N | |
108829 | |
C1=CC(=C(C=C1Br)Cl)C(=O)O |
C7H4BrClO2 | |
MFCD00040903 | |
benzoic acid, 4-bromo-2-chloro, 4-bromo-2-chlorobenzoicacid, 2-chloro-4-bromobenzoic acid, 4-bromo-2-chloro-benzoic acid, rarechem al bo 2371, attercop-chm at111793, zlchem 410, pubchem3586, acmc-1b03i, ksc274c2d | |
4-bromo-2-chlorobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 261-911-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only