missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-chlorobenzenesulfonyl chloride, 97%, ACROS Organics™

Chemical Identifiers

CAS 351003-52-6
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H3BrCl2O2S
Linear Formula BrC6H3ClSO2Cl
Formula Weight 289.96
MDL Number MFCD03094630
Chemical Name or Material 4-Bromo-2-chlorobenzenesulfonyl chloride, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC371430050 SDS Thermo Fisher Scientific
AC371430050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC371430010 SDS Thermo Fisher Scientific
AC371430010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

351003-52-6
100.0
C6H3BrCl2O2S
289.96
4-Bromo-2-chlorobenzenesulfonyl chloride, 97%
96.0
97%
BrC6H3ClSO2Cl
MFCD03094630
≥96.0%

Specifications

351003-52-6
96.0
Glass bottle
75° to 80°C
Crystals or Chunks
C6H3BrCl2O2S
289.96
4-Bromo-2-chlorobenzenesulfonyl chloride, 97%
Authentic
100.0
Yellow
5g
97%
BrC6H3ClSO2Cl
MFCD03094630
≥96.0%

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger