Please login to your online account to display your discounted pricing

4-Bromo-2-chlorobenzenesulfonyl chloride, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC371430010

 View more versions of this product

Catalog No. AC371430010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 289.96
Physical Form Crystals or Chunks
Assay ≥96.0%
Quantity 1g
Color Yellow
Melting Point 75° to 80°C
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
CAS 351003-52-6
Molecular Formula C6H3BrCl2O2S
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 4-Bromo-2-chlorobenzenesulfonyl chloride
Name Note 97%
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula BrC6H3ClSO2Cl
MDL Number MFCD03094630

Specifications

Chemical Name or Material 4-Bromo-2-chlorobenzenesulfonyl chloride
Name Note 97%
CAS 351003-52-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H3BrCl2O2S
Linear Formula BrC6H3ClSO2Cl
Formula Weight 289.96
MDL Number MFCD03094630
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents