Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-chlorobenzenesulfonamide, 97%

$84.82 - $328.47

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-chlorobenzenesulfonamide
CAS 351003-59-3
Molecular Formula C6H5BrClNO2S
Formula Weight 270.53
MDL Number MFCD03427209
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1971906

  SDS
alfa aesar™
L1971906
5g Each for $328.47

AAL1971903

  SDS
alfa aesar™
L1971903
1g Each for $84.82
Description & Specifications

Specifications

Formula Weight 270.53
Chemical Name or Material 4-Bromo-2-chlorobenzenesulfonamide
CAS 351003-59-3
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C6H5BrClNO2S
MDL Number MFCD03427209
Melting Point 172° to 176°C
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.