Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-chlorobenzaldehyde, 95%

$135.61 - $308.89

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-chlorobenzaldehyde
CAS 158435-41-7
Molecular Formula C7H4BrClO
Formula Weight 219.47
MDL Number MFCD08741413
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2741803

  SDS
alfa aesar™
H2741803
1g Each for $308.89

AAH27418MD

  SDS
alfa aesar™
H27418MD
250mg Each for $135.61
Description & Specifications

Specifications

Formula Weight 219.47
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD08741413
CAS 158435-41-7
Assay 95%
Molecular Formula C7H4BrClO
Health Hazard 1 Warning
Quantity 1g
Melting Point 74° to 76°C
Chemical Name or Material 4-Bromo-2-chlorobenzaldehyde
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.