missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-chloroaniline, 99%, ACROS Organics™

Manufacturer:  Acros Organics 167480500

 View more versions of this product

Catalog No. AC167480500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Bromo-2-chloroaniline
Glass bottle
67°C to 70°C
99%
98.5
99%
BrC6H3(Cl)NH2
12, 652
INMZDDDQLHKGPF-UHFFFAOYSA-N
4-bromo-2-chloroaniline
610169
Crystalline Powder
Authentic
Brown to Yellow
50g
38762-41-3
100.0
C6H5BrClN
MFCD00007660
2-chloro-4-bromoaniline, 4-bromo-2-chloro-phenylamine, benzenamine, 4-bromo-2-chloro, 4-bromo-2-chloro-aniline, 4-bromo-2-chlorophenylamine, 4-bromo-6-chloroaniline, 4-bromo-2-chlorobenzenamine, zlchem 546, pubchem3578, 2-chloro4-bromoaniline
C1=CC(=C(C=C1Br)Cl)N
206.467
206.47
≥98.5%

Chemical Identifiers

38762-41-3
206.467
INMZDDDQLHKGPF-UHFFFAOYSA-N
610169
C1=CC(=C(C=C1Br)Cl)N
C6H5BrClN
MFCD00007660
2-chloro-4-bromoaniline, 4-bromo-2-chloro-phenylamine, benzenamine, 4-bromo-2-chloro, 4-bromo-2-chloro-aniline, 4-bromo-2-chlorophenylamine, 4-bromo-6-chloroaniline, 4-bromo-2-chlorobenzenamine, zlchem 546, pubchem3578, 2-chloro4-bromoaniline
4-bromo-2-chloroaniline
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 254-118-4

SDS Documents