Please login to your online account to display your discounted pricing

4-Bromo-2-chloroaniline, 99%, ACROS Organics™


Quantity 10g

Manufacturer: thermo fisher scientific  167480100

 View more versions of this product

Catalog No. AC167480100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 206.47
Assay ≥98.5%
Quantity 10g
Color Slightly yellow to light brown
Beilstein 12, 652
EINECS Number 254-118-4
Melting Point 67° to 70°C
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula BrC6H3(Cl)NH2
Chemical Name or Material 4-Bromo-2-chloroaniline
CAS Min % 98.5
Name Note 99%
Assay Percent Range 99%
Molecular Formula C6H5BrClN
MDL Number MFCD00007660
CAS 38762-41-3
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material 4-Bromo-2-chloroaniline
Name Note 99%
CAS 38762-41-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H5BrClN
Linear Formula BrC6H3(Cl)NH2
Formula Weight 206.47
MDL Number MFCD00007660
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.

SDS & Safety Documents