Please login to your online account to display your discounted pricing

4-Bromo-2-chloro-6-methylphenol, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SB01216DA

 View more versions of this product

Catalog No. SB01216DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 221.48
Quantity 1g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
CAS 7530-27-0
Molecular Formula C7H6BrClO
Chemical Name or Material 4-Bromo-2-chloro-6-methylphenol
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00002167
Linear Formula CH3C6H2(OH)(Cl)Br
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Assay Percent Range 97%

Specifications

Chemical Name or Material 4-Bromo-2-chloro-6-methylphenol
CAS 7530-27-0
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H6BrClO
Linear Formula CH3C6H2(OH)(Cl)Br
Formula Weight 221.48
MDL Number MFCD00002167
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents