missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-chloro-6-methylaniline, 98%

Chemical Identifiers

CAS 30273-42-8
Molecular Formula C7H7BrClN
Formula Weight 220.5
MDL Number MFCD00041432
UN Number UN2811
Beilstein 2717133
Synonym 4-Bromo-6-chloro-o-toluidine
Chemical Name or Material 4-Bromo-2-chloro-6-methylaniline
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1259006 SDS Alfa Aesar™
L1259006
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAL1259014 SDS Alfa Aesar™
L1259014
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

30273-42-8
220.5
UN2811
4-Bromo-6-chloro-o-toluidine
C7H7BrClN
MFCD00041432
2717133
4-Bromo-2-chloro-6-methylaniline

Specifications

30273-42-8
220.5
UN2811
2717133
5g
98%
C7H7BrClN
MFCD00041432
37° to 40°C
4-Bromo-6-chloro-o-toluidine
4-Bromo-2-chloro-6-methylaniline

Safety and Handling

GHS H Statement
H331-H373-H302-H312
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P261-P280-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.