Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-chloro-6-fluoroanisole, 97%

$156.70 - $473.12

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-chloro-6-fluoroanisole
CAS 261762-34-9
Molecular Formula C7H5BrClFO
Formula Weight 239.47
MDL Number MFCD01631477
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2663503

  SDS
alfa aesar™
H2663503
1g Each for $156.70

AAH2663506

  SDS
alfa aesar™
H2663506
5g Each for $473.12
Description & Specifications

Specifications

Formula Weight 239.47
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
CAS 261762-34-9
Molecular Formula C7H5BrClFO
Quantity 1g
Chemical Name or Material 4-Bromo-2-chloro-6-fluoroanisole
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
MDL Number MFCD01631477
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.