missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-chloro-5-fluorobenzoic acid, 98%

$31.30 - $558.24

Chemical Identifiers

CAS 177480-81-8
Molecular Formula C7H3BrClFO2
Molecular Weight (g/mol) 253.451
MDL Number MFCD00236589
InChI Key TZFKKLGCVKORLS-UHFFFAOYSA-N
Synonym 4-bromo-2-chloro-5-fluoro-benzoic acid, benzoic acid, 4-bromo-2-chloro-5-fluoro, pubchem1361, acmc-1c1y0, 4-bromo-2-chloro-5-fluorobenzoicacid, 2-chloro-4-bromo-5-fluorobenzoic acid
PubChem CID 2736225
IUPAC Name 4-bromo-2-chloro-5-fluorobenzoic acid
SMILES C1=C(C(=CC(=C1F)Br)Cl)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6396503 View Documents Alfa Aesar™
H6396503
1g Each for $31.30
 
AAH6396506 View Documents Alfa Aesar™
H6396506
5g Each for $130.62
 
AAH6396514 View Documents Alfa Aesar™
H6396514
25g Each for $558.24
 
Specifications

Chemical Identifiers

177480-81-8
253.451
TZFKKLGCVKORLS-UHFFFAOYSA-N
2736225
C1=C(C(=CC(=C1F)Br)Cl)C(=O)O
C7H3BrClFO2
MFCD00236589
4-bromo-2-chloro-5-fluoro-benzoic acid, benzoic acid, 4-bromo-2-chloro-5-fluoro, pubchem1361, acmc-1c1y0, 4-bromo-2-chloro-5-fluorobenzoicacid, 2-chloro-4-bromo-5-fluorobenzoic acid
4-bromo-2-chloro-5-fluorobenzoic acid

Specifications

White
177480-81-8
MFCD00236589
TZFKKLGCVKORLS-UHFFFAOYSA-N
4-bromo-2-chloro-5-fluorobenzoic acid
2736225
Solid
4-Bromo-2-chloro-5-fluorobenzoic acid
1g
C7H3BrClFO2
4-bromo-2-chloro-5-fluoro-benzoic acid, benzoic acid, 4-bromo-2-chloro-5-fluoro, pubchem1361, acmc-1c1y0, 4-bromo-2-chloro-5-fluorobenzoicacid, 2-chloro-4-bromo-5-fluorobenzoic acid
C1=C(C(=CC(=C1F)Br)Cl)C(=O)O
253.451
253.45
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning