Learn More
4-Bromo-2,6-dimethylaniline, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 185190250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Bromo-2, 6-dimethylaniline | |
24596-19-8 | |
100.0 | |
Authentic | |
Glass bottle | |
BrC6H2(CH3)2NH2 | |
25g | |
4-bromo-2,6-xylidine, benzenamine, 4-bromo-2,6-dimethyl, 4-bromo-2,6-dimethyl-phenylamine, 2-amino-5-bromo-m-xylene, 2,6-dimethyl-4-bromoaniline, 4-bromo-2,6-dimethylbenzenamine, 4-bromo-2,6-dimethylphenylamine, 4-bromo-2,6-dimethyl-aniline, 4-bromo-2,6-dimethylaniline, 4-bromo-2,6-dimethyl-benzenamine | |
QGLAYJCJLHNIGJ-UHFFFAOYSA-N | |
4-bromo-2,6-dimethylaniline | |
90549 | |
98% |
98% | |
97.5 | |
>110°C | |
97.5% min. (GC) | |
C8H10BrN | |
MFCD00007826 | |
12,111 | |
Solubility in water: insoluble | |
CC1=CC(Br)=CC(C)=C1N | |
200.08 | |
200.08 |
Chemical Identifiers
24596-19-8 | |
200.08 | |
QGLAYJCJLHNIGJ-UHFFFAOYSA-N | |
90549 | |
CC1=CC(Br)=CC(C)=C1N |
C8H10BrN | |
MFCD00007826 | |
4-bromo-2,6-xylidine, benzenamine, 4-bromo-2,6-dimethyl, 4-bromo-2,6-dimethyl-phenylamine, 2-amino-5-bromo-m-xylene, 2,6-dimethyl-4-bromoaniline, 4-bromo-2,6-dimethylbenzenamine, 4-bromo-2,6-dimethylphenylamine, 4-bromo-2,6-dimethyl-aniline, 4-bromo-2,6-dimethylaniline, 4-bromo-2,6-dimethyl-benzenamine | |
4-bromo-2,6-dimethylaniline |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a P
GHS Signal Word: Warning
EINECSNumber : 246-337-9
RUO â Research Use Only