missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromo-2,6-dimethylaniline, 98%, ACROS Organics™
Chemical Identifiers
CAS | 24596-19-8 |
---|---|
Molecular Formula | C8H10BrN |
Molecular Weight (g/mol) | 200.08 |
MDL Number | MFCD00007826 |
InChI Key | QGLAYJCJLHNIGJ-UHFFFAOYSA-N |
Synonym | 2,6-dimethyl-4-bromoaniline, 2-amino-5-bromo-m-xylene, 4-bromo-2,6-dimethyl-aniline, 4-bromo-2,6-dimethyl-benzenamine, 4-bromo-2,6-dimethyl-phenylamine, 4-bromo-2,6-dimethylaniline, 4-bromo-2,6-dimethylbenzenamine, 4-bromo-2,6-dimethylphenylamine, 4-bromo-2,6-xylidine, benzenamine, 4-bromo-2,6-dimethyl |
PubChem CID | 90549 |
IUPAC Name | 4-bromo-2,6-dimethylaniline |
SMILES | CC1=CC(=CC(=C1N)C)Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC185190050
|
Thermo Fisher Scientific
185190050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC185190250
|
Thermo Fisher Scientific
185190250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC185192500
|
Thermo Fisher Scientific
AC185192500 |
250g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
24596-19-8 | |
200.08 | |
QGLAYJCJLHNIGJ-UHFFFAOYSA-N | |
90549 | |
CC1=CC(=CC(=C1N)C)Br |
C8H10BrN | |
MFCD00007826 | |
2,6-dimethyl-4-bromoaniline, 2-amino-5-bromo-m-xylene, 4-bromo-2,6-dimethyl-aniline, 4-bromo-2,6-dimethyl-benzenamine, 4-bromo-2,6-dimethyl-phenylamine, 4-bromo-2,6-dimethylaniline, 4-bromo-2,6-dimethylbenzenamine, 4-bromo-2,6-dimethylphenylamine, 4-bromo-2,6-xylidine, benzenamine, 4-bromo-2,6-dimethyl | |
4-bromo-2,6-dimethylaniline |
Specifications
>110°C | |
Glass bottle | |
48.0°C to 52.0°C | |
24596-19-8 | |
100.0 | |
C8H10BrN | |
MFCD00007826 | |
2,6-dimethyl-4-bromoaniline, 2-amino-5-bromo-m-xylene, 4-bromo-2,6-dimethyl-aniline, 4-bromo-2,6-dimethyl-benzenamine, 4-bromo-2,6-dimethyl-phenylamine, 4-bromo-2,6-dimethylaniline, 4-bromo-2,6-dimethylbenzenamine, 4-bromo-2,6-dimethylphenylamine, 4-bromo-2,6-xylidine, benzenamine, 4-bromo-2,6-dimethyl | |
QGLAYJCJLHNIGJ-UHFFFAOYSA-N | |
4-bromo-2,6-dimethylaniline | |
90549 | |
Liquid | |
4-Bromo-2, 6-dimethylaniline, 98% |
Authentic | |
Amber to Yellow | |
5g | |
97.5 | |
97.5% min. (GC) | |
BrC6H2(CH3)2NH2 | |
12,111 | |
Solubility in water: insoluble | |
CC1=CC(=CC(=C1N)C)Br | |
200.08 | |
200.08 | |
98% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a P
GHS Signal Word: Warning
EINECSNumber : 246-337-9