Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2,6-difluorophenol, 97%

$264.76 - $264.76

Chemical Identifiers

Chemical Name or Material 4-Bromo-2,6-difluorophenol
CAS 104197-13-9
Molecular Formula C6H3BrF2O
Formula Weight 208.99
MDL Number MFCD00236213
Beilstein 8543473
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2032906

 
alfa aesar™
B2032906
5g Each for $264.76
Description & Specifications

Specifications

Formula Weight 208.99
Chemical Name or Material 4-Bromo-2,6-difluorophenol
Assay 97%
Molecular Formula C6H3BrF2O
MDL Number MFCD00236213
CAS 104197-13-9
Beilstein 8543473
Melting Point ≈50°C
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.