missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromo-2,6-difluorobenzyl alcohol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6162303
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Bromo-2,6-difluorobenzyl alcohol | |
76°C to 81°C | |
MFCD03094461 | |
4-bromo-2,6-difluorobenzyl alcohol, 4-bromo-2,6-difluorophenyl methanol, 4-bromo-2,6-difluorobenzylalcohol, benzenemethanol, 4-bromo-2,6-difluoro, 4-bromo-2,6-difluoro-benzenemethanol, 2,6-difluoro-4-bromobenzyl alcohol, 4-bromo-2,6-difluoro-phenyl methanol, 2,6-difluoro-4-bromo benzyl alcohol, pubchem4882, acmc-209dox | |
C1=C(C=C(C(=C1F)CO)F)Br | |
223.017 | |
223.01 | |
Solid |
162744-59-4 | |
C7H5BrF2O | |
1 g | |
LSRHFWSNUFIKER-UHFFFAOYSA-N | |
(4-bromo-2,6-difluorophenyl)methanol | |
2773303 | |
97% |
Chemical Identifiers
162744-59-4 | |
223.017 | |
LSRHFWSNUFIKER-UHFFFAOYSA-N | |
2773303 | |
C1=C(C=C(C(=C1F)CO)F)Br |
C7H5BrF2O | |
MFCD03094461 | |
4-bromo-2,6-difluorobenzyl alcohol, 4-bromo-2,6-difluorophenyl methanol, 4-bromo-2,6-difluorobenzylalcohol, benzenemethanol, 4-bromo-2,6-difluoro, 4-bromo-2,6-difluoro-benzenemethanol, 2,6-difluoro-4-bromobenzyl alcohol, 4-bromo-2,6-difluoro-phenyl methanol, 2,6-difluoro-4-bromo benzyl alcohol, pubchem4882, acmc-209dox | |
(4-bromo-2,6-difluorophenyl)methanol |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only