missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2,6-difluorobenzonitrile, TCI America™

$32.08 - $91.67

Chemical Identifiers

CAS 123843-67-4
Molecular Formula C7H2BrF2N
Molecular Weight (g/mol) 218.001
MDL Number MFCD01310981
InChI Key TZHQWUAOIWRFSW-UHFFFAOYSA-N
Synonym 4-bromo-2,6-difluoro-benzonitrile, 2,6-difluoro-4-bromobenzonitrile, benzonitrile, 4-bromo-2,6-difluoro, 4-bromo-2,6-difluorobenzenecarbonitrile, pubchem4699, acmc-1bymk, ksc513m5d, 4-bromo-2,6-difluoro-bezonitrile, 2,6-difluoro-4-bromo-benzonitrile
PubChem CID 2756916
IUPAC Name 4-bromo-2,6-difluorobenzonitrile
SMILES C1=C(C=C(C(=C1F)C#N)F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B40211G View Documents TCI America
B40211G
1g Each for $32.08
 
B40215G View Documents TCI America
B40215G
5g Each for $91.67
 
Specifications

Chemical Identifiers

123843-67-4
218.001
TZHQWUAOIWRFSW-UHFFFAOYSA-N
2756916
C1=C(C=C(C(=C1F)C#N)F)Br
C7H2BrF2N
MFCD01310981
4-bromo-2,6-difluoro-benzonitrile, 2,6-difluoro-4-bromobenzonitrile, benzonitrile, 4-bromo-2,6-difluoro, 4-bromo-2,6-difluorobenzenecarbonitrile, pubchem4699, acmc-1bymk, ksc513m5d, 4-bromo-2,6-difluoro-bezonitrile, 2,6-difluoro-4-bromo-benzonitrile
4-bromo-2,6-difluorobenzonitrile

Specifications

White
1g
C7H2BrF2N
3439
TZHQWUAOIWRFSW-UHFFFAOYSA-N
4-bromo-2,6-difluorobenzonitrile
2756916
Crystal-Powder at 20°C
4-Bromo-2,6-difluorobenzonitrile
81°C
123843-67-4
MFCD01310981
4-bromo-2,6-difluoro-benzonitrile, 2,6-difluoro-4-bromobenzonitrile, benzonitrile, 4-bromo-2,6-difluoro, 4-bromo-2,6-difluorobenzenecarbonitrile, pubchem4699, acmc-1bymk, ksc513m5d, 4-bromo-2,6-difluoro-bezonitrile, 2,6-difluoro-4-bromo-benzonitrile
C1=C(C=C(C(=C1F)C#N)F)Br
218.001
218.00
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No