Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2,6-difluorobenzoic acid, 98%

$49.26 - $138.08

Chemical Identifiers

Chemical Name or Material 4-Bromo-2,6-difluorobenzoic acid
CAS 183065-68-1
Molecular Formula C7H3BrF2O2
Formula Weight 237
MDL Number MFCD03094085
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2609003

 
alfa aesar™
H2609003
1g Each for $138.08

AAH26090MD

 
alfa aesar™
H26090MD
250mg Each for $49.26
Description & Specifications

Specifications

Formula Weight 237
Chemical Name or Material 4-Bromo-2,6-difluorobenzoic acid
Assay 98%
Molecular Formula C7H3BrF2O2
MDL Number MFCD03094085
CAS 183065-68-1
Melting Point 202° to 204°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.