missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2,6-difluorobenzaldehyde, TCI America™

$13.75 - $48.89

Chemical Identifiers

CAS 537013-51-7
Molecular Formula C7H3BrF2O
Molecular Weight (g/mol) 221.001
MDL Number MFCD03094459
InChI Key CZGVAISJIQNQEJ-UHFFFAOYSA-N
Synonym 4-bromo-2,6-difluorobenzylaldehyde, 4-bromo-2,6-difluoro-benzaldehyde, benzaldehyde, 4-bromo-2,6-difluoro, 2,6-difluoro-4-bromobenzaldehyde, 2,6-difluoro-4-bromo benzaldehyde, pubchem4165, acmc-209l9s, ksc911q5f, 4-bromo 2,6-difluorobenzaldehyde, 2,6-difluoro-4-bromo-benzaldehyde
PubChem CID 2773290
IUPAC Name 4-bromo-2,6-difluorobenzaldehyde
SMILES C1=C(C=C(C(=C1F)C=O)F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B3446100MG View Documents TCI America
B3446100MG
100mg Each for $13.75
 
B34461G View Documents TCI America
B34461G
1g Each for $48.89
 
Specifications

Chemical Identifiers

537013-51-7
221.001
CZGVAISJIQNQEJ-UHFFFAOYSA-N
2773290
C1=C(C=C(C(=C1F)C=O)F)Br
C7H3BrF2O
MFCD03094459
4-bromo-2,6-difluorobenzylaldehyde, 4-bromo-2,6-difluoro-benzaldehyde, benzaldehyde, 4-bromo-2,6-difluoro, 2,6-difluoro-4-bromobenzaldehyde, 2,6-difluoro-4-bromo benzaldehyde, pubchem4165, acmc-209l9s, ksc911q5f, 4-bromo 2,6-difluorobenzaldehyde, 2,6-difluoro-4-bromo-benzaldehyde
4-bromo-2,6-difluorobenzaldehyde

Specifications

Undesignated
100mg
C7H3BrF2O
4-bromo-2,6-difluorobenzylaldehyde, 4-bromo-2,6-difluoro-benzaldehyde, benzaldehyde, 4-bromo-2,6-difluoro, 2,6-difluoro-4-bromobenzaldehyde, 2,6-difluoro-4-bromo benzaldehyde, pubchem4165, acmc-209l9s, ksc911q5f, 4-bromo 2,6-difluorobenzaldehyde, 2,6-difluoro-4-bromo-benzaldehyde
C1=C(C=C(C(=C1F)C=O)F)Br
221.001
221.00
≥98.0% (GC)
77°C
537013-51-7
MFCD03094459
CZGVAISJIQNQEJ-UHFFFAOYSA-N
4-bromo-2,6-difluorobenzaldehyde
2773290
Crystal-Powder at 20°C
4-Bromo-2,6-difluorobenzaldehyde
Safety and Handling

Safety and Handling

TSCA : No