Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2,6-difluorobenzaldehyde, 95%

$262.18 - $522.85

Chemical Identifiers

Chemical Name or Material 4-Bromo-2,6-difluorobenzaldehyde
CAS 537013-51-7
Molecular Formula C7H3BrF2O
Formula Weight 221
MDL Number MFCD03094459
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2773703

  SDS
alfa aesar™
H2773703
1g Each for $522.85

AAH27737MD

  SDS
alfa aesar™
H27737MD
250mg Each for $262.18
Description & Specifications

Specifications

Formula Weight 221
Quantity 1g
Melting Point 76° to 79°C
MDL Number MFCD03094459
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Chemical Name or Material 4-Bromo-2,6-difluorobenzaldehyde
CAS 537013-51-7
Molecular Formula C7H3BrF2O
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.