Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2,6-difluoroanisole, 97+%

$88.15 - $323.96

Chemical Identifiers

Chemical Name or Material 4-Bromo-2,6-difluoroanisole
CAS 104197-14-0
Molecular Formula C7H5BrF2O
Formula Weight 223.02
MDL Number MFCD01631351
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2428203

  SDS
alfa aesar™
B2428203
1g Each for $88.15

AAB2428206

  SDS
alfa aesar™
B2428206
5g Each for $323.96
Description & Specifications

Specifications

Formula Weight 223.02
MDL Number MFCD01631351
Chemical Name or Material 4-Bromo-2,6-difluoroanisole
Assay ≥97%
Molecular Formula C7H5BrF2O
CAS 104197-14-0
Boiling Point 100°C (15mmHg)
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
t is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.