missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2,6-difluoroaniline, 98%, ACROS Organics™

Chemical Identifiers

CAS 67567-26-4
Molecular Formula C6H4BrF2N
Molecular Weight (g/mol) 208.01
InChI Key BFQSQUAVMNHOEF-UHFFFAOYSA-N
Synonym 2,6-difluoro-4-bromoaniline, 4-bromo-2,6-difluoro-aniline, 4-bromo-2,6-difluoro-phenylamine, 4-bromo-2,6-difluorobenzenamine, 4-bromo-2,6-difluorophenyl amine, 4-bromo-2,6-difluorophenylamine, acmc-1begp, benzenamine, 4-bromo-2,6-difluoro, buttpark 22\07-66, pubchem2929
PubChem CID 610191
IUPAC Name 4-bromo-2,6-difluoroaniline
SMILES C1=C(C=C(C(=C1F)N)F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC442220010
SDS View Documents
Thermo Fisher Scientific
AC442220010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC442220050
SDS View Documents
Thermo Fisher Scientific
AC442220050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

67567-26-4
208.01
2,6-difluoro-4-bromoaniline, 4-bromo-2,6-difluoro-aniline, 4-bromo-2,6-difluoro-phenylamine, 4-bromo-2,6-difluorobenzenamine, 4-bromo-2,6-difluorophenyl amine, 4-bromo-2,6-difluorophenylamine, acmc-1begp, benzenamine, 4-bromo-2,6-difluoro, buttpark 22\07-66, pubchem2929
4-bromo-2,6-difluoroaniline
C6H4BrF2N
BFQSQUAVMNHOEF-UHFFFAOYSA-N
610191
C1=C(C=C(C(=C1F)N)F)Br

Specifications

Authentic
Brown
1g
97.5
97.5% min. (GC)
2,6-difluoro-4-bromoaniline, 4-bromo-2,6-difluoro-aniline, 4-bromo-2,6-difluoro-phenylamine, 4-bromo-2,6-difluorobenzenamine, 4-bromo-2,6-difluorophenyl amine, 4-bromo-2,6-difluorophenylamine, acmc-1begp, benzenamine, 4-bromo-2,6-difluoro, buttpark 22\07-66, pubchem2929
C1=C(C=C(C(=C1F)N)F)Br
208.01
208.01
98%
Glass Bottle
67°C to 68°C
67567-26-4
100.0
C6H4BrF2N
BFQSQUAVMNHOEF-UHFFFAOYSA-N
4-bromo-2,6-difluoroaniline
610191
Crystalline Powder
4-Bromo-2, 6-difluoroaniline
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning