Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2,6-difluoroaniline, 98%

$64.50 - $213.96

Chemical Identifiers

Chemical Name or Material 4-Bromo-2,6-difluoroaniline
CAS 67567-26-4
Molecular Formula C6H4BrF2N
Formula Weight 208.01
MDL Number MFCD00013389
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2054903

 
alfa aesar™
B2054903
1g Each for $64.50

AAB2054906

 
alfa aesar™
B2054906
5g Each for $213.96
Description & Specifications

Specifications

Formula Weight 208.01
Chemical Name or Material 4-Bromo-2,6-difluoroaniline
Assay 98%
Molecular Formula C6H4BrF2N
MDL Number MFCD00013389
CAS 67567-26-4
Melting Point 63° to 65°C
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.