Learn More
4-Bromo-2,6-dichloroaniline, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1488414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Bromo-2,6-dichloroaniline | |
85°C to 88°C | |
MFCD00025162 | |
2089687 | |
NPQBZKNXJZARBJ-UHFFFAOYSA-N | |
4-bromo-2,6-dichloroaniline | |
69680 | |
98% |
697-88-1 | |
C6H4BrCl2N | |
25 g | |
2,6-dichloro-4-bromoaniline, 4-bromo-2,6-dichlorophenyl amine, 4-bromo-2,6-dichlorophenylamine, 4-bromo-2,6-dichloro-benzenamine, benzenamine, 4-bromo-2,6-dichloro, 4-brom-2,6-dichloranilin, pubchem3606, acmc-1ay6t, ksc495a6h, 4-bromo-2,6-dichloro aniline | |
C1=C(C=C(C(=C1Cl)N)Cl)Br | |
240.909 | |
240.92 |
Chemical Identifiers
697-88-1 | |
240.909 | |
NPQBZKNXJZARBJ-UHFFFAOYSA-N | |
69680 | |
C1=C(C=C(C(=C1Cl)N)Cl)Br |
C6H4BrCl2N | |
MFCD00025162 | |
2,6-dichloro-4-bromoaniline, 4-bromo-2,6-dichlorophenyl amine, 4-bromo-2,6-dichlorophenylamine, 4-bromo-2,6-dichloro-benzenamine, benzenamine, 4-bromo-2,6-dichloro, 4-brom-2,6-dichloranilin, pubchem3606, acmc-1ay6t, ksc495a6h, 4-bromo-2,6-dichloro aniline | |
4-bromo-2,6-dichloroaniline |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 211-809-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only