missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2,6-dichloroaniline, 98%

Manufacturer:  Alfa Aesar™ A1488406

 View more versions of this product

Catalog No. AAA1488406



Specifications

Specifications

4-Bromo-2,6-dichloroaniline
5g
C6H4BrCl2N
2089687
NPQBZKNXJZARBJ-UHFFFAOYSA-N
4-bromo-2,6-dichloroaniline
69680
98%
85°C to 88°C
697-88-1
MFCD00025162
2,6-dichloro-4-bromoaniline, 4-bromo-2,6-dichlorophenyl amine, 4-bromo-2,6-dichlorophenylamine, 4-bromo-2,6-dichloro-benzenamine, benzenamine, 4-bromo-2,6-dichloro, 4-brom-2,6-dichloranilin, pubchem3606, acmc-1ay6t, ksc495a6h, 4-bromo-2,6-dichloro aniline
C1=C(C=C(C(=C1Cl)N)Cl)Br
240.909
240.92

Chemical Identifiers

697-88-1
240.909
NPQBZKNXJZARBJ-UHFFFAOYSA-N
69680
C1=C(C=C(C(=C1Cl)N)Cl)Br
C6H4BrCl2N
MFCD00025162
2,6-dichloro-4-bromoaniline, 4-bromo-2,6-dichlorophenyl amine, 4-bromo-2,6-dichlorophenylamine, 4-bromo-2,6-dichloro-benzenamine, benzenamine, 4-bromo-2,6-dichloro, 4-brom-2,6-dichloranilin, pubchem3606, acmc-1ay6t, ksc495a6h, 4-bromo-2,6-dichloro aniline
4-bromo-2,6-dichloroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents