Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-(5-isoxazolyl)phenol, 97%

$32.54 - $322.45

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-(5-isoxazolyl)phenol
CAS 213690-27-8
Molecular Formula C9H6BrNO2
Formula Weight 240.05
MDL Number MFCD00799493
Synonym 5-(5-Bromo-2-hydroxyphenyl)isoxazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2755703

  SDS
alfa aesar™
H2755703
1g Each for $32.54

AAH2755706

  SDS
alfa aesar™
H2755706
5g Each for $99.30

AAH2755714

  SDS
alfa aesar™
H2755714
25g Each for $322.45
Description & Specifications

Specifications

Formula Weight 240.05
CAS 213690-27-8
Synonym 5-(5-Bromo-2-hydroxyphenyl)isoxazole
Molecular Formula C9H6BrNO2
Assay 97%
MDL Number MFCD00799493
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Bromo-2-(5-isoxazolyl)phenol
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 207° to 212°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.