Please login to your online account to display your discounted pricing

4-Bromo-2,5-difluorobenzenesulfonyl chloride, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC399880050

 View more versions of this product

Catalog No. AC399880050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder or Crystals
Assay ≥97.5% (GC)
Formula Weight 291.49
Melting Point 32° to 38°C
Quantity 5g
Color White to yellow or orange, may darken on storage
CAS Max % 100.0
Packaging Glass bottle
Linear Formula F2C6H2(Br)SO2Cl
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Molecular Formula C6H2BrClF2O2S
CAS 207974-14-9
CAS Min % 97.5
Chemical Name or Material 4-Bromo-2, 5-difluorobenzenesulfonyl chloride
Assay Percent Range 98%
MDL Number MFCD00042181

Specifications

Chemical Name or Material 4-Bromo-2, 5-difluorobenzenesulfonyl chloride
CAS 207974-14-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C6H2BrClF2O2S
Linear Formula F2C6H2(Br)SO2Cl
Formula Weight 291.49
MDL Number MFCD00042181
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

SDS & Safety Documents