missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2,5-difluorobenzenesulfonyl chloride, 98%, ACROS Organics™

Chemical Identifiers

CAS 207974-14-9
Molecular Formula C6H2BrClF2O2S
Molecular Weight (g/mol) 291.49
MDL Number MFCD00042181
InChI Key SBMKFWMFNIEPDN-UHFFFAOYSA-N
Synonym 4-bromo-2,5-difluorobenzene-1-sulfonyl chloride, 4-bromo-2,5-difluorobenzene-1-sulfonylchloride, 4-bromo-2,5-difluorobenzenesulfonylchloride, 4-bromo-2,5-difluorobenzenesulphonyl chloride, 4-bromo-2,5-difluorobenzenesulphonylchloride, 4-bromo-2,5-difluorophenyl chlorosulfone, 4-bromo-3,6-difluoro-benzenesulfonylchloride, acmc-209feg, benzenesulfonyl chloride, 4-bromo-2,5-difluoro, pubchem5058
PubChem CID 574590
IUPAC Name 4-bromo-2,5-difluorobenzenesulfonyl chloride
SMILES C1=C(C(=CC(=C1S(=O)(=O)Cl)F)Br)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC399880010
SDS View Documents
Thermo Fisher Scientific
AC399880010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC399880050
SDS View Documents
Thermo Fisher Scientific
AC399880050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

207974-14-9
291.49
SBMKFWMFNIEPDN-UHFFFAOYSA-N
574590
C1=C(C(=CC(=C1S(=O)(=O)Cl)F)Br)F
C6H2BrClF2O2S
MFCD00042181
4-bromo-2,5-difluorobenzene-1-sulfonyl chloride, 4-bromo-2,5-difluorobenzene-1-sulfonylchloride, 4-bromo-2,5-difluorobenzenesulfonylchloride, 4-bromo-2,5-difluorobenzenesulphonyl chloride, 4-bromo-2,5-difluorobenzenesulphonylchloride, 4-bromo-2,5-difluorophenyl chlorosulfone, 4-bromo-3,6-difluoro-benzenesulfonylchloride, acmc-209feg, benzenesulfonyl chloride, 4-bromo-2,5-difluoro, pubchem5058
4-bromo-2,5-difluorobenzenesulfonyl chloride

Specifications

Authentic
Orange or White to Yellow
1g
97.5
97.5 % min. (GC)
F2C6H2(Br)SO2Cl
4-bromo-2,5-difluorobenzene-1-sulfonyl chloride, 4-bromo-2,5-difluorobenzene-1-sulfonylchloride, 4-bromo-2,5-difluorobenzenesulfonylchloride, 4-bromo-2,5-difluorobenzenesulphonyl chloride, 4-bromo-2,5-difluorobenzenesulphonylchloride, 4-bromo-2,5-difluorophenyl chlorosulfone, 4-bromo-3,6-difluoro-benzenesulfonylchloride, acmc-209feg, benzenesulfonyl chloride, 4-bromo-2,5-difluoro, pubchem5058
C1=C(C(=CC(=C1S(=O)(=O)Cl)F)Br)F
291.49
291.49
98%
Glass bottle
37°C to 39°C
207974-14-9
100.0
C6H2BrClF2O2S
MFCD00042181
SBMKFWMFNIEPDN-UHFFFAOYSA-N
4-bromo-2,5-difluorobenzenesulfonyl chloride
574590
Crystalline Powder or Crystals
4-Bromo-2, 5-difluorobenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger