Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2,5-difluorobenzenesulfonamide, 98+%

$43.41 - $43.41

Chemical Identifiers

Chemical Name or Material 4-Bromo-2,5-difluorobenzenesulfonamide
CAS 214209-98-0
Molecular Formula C6H4BrF2NO2S
Formula Weight 272.07
MDL Number MFCD01569463
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2601003

  SDS
alfa aesar™
H2601003
1g Each for $43.41
Description & Specifications

Specifications

Formula Weight 272.07
Chemical Name or Material 4-Bromo-2,5-difluorobenzenesulfonamide
Assay ≥98%
Molecular Formula C6H4BrF2NO2S
MDL Number MFCD01569463
CAS 214209-98-0
Melting Point 164° to 166°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.