Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2,5-difluoroanisole, 99%

$48.97 - $177.80

Chemical Identifiers

Chemical Name or Material 4-Bromo-2,5-difluoroanisole
CAS 202865-60-9
Molecular Formula C7H5BrF2O
Formula Weight 223.02
MDL Number MFCD00143258
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1737406

  SDS
alfa aesar™
A1737406
5g Each for $48.97

AAA1737414

  SDS
alfa aesar™
A1737414
25g Each for $177.80
Description & Specifications

Specifications

Formula Weight 223.02
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material 4-Bromo-2,5-difluoroanisole
Quantity 5g
Melting Point 61° to 64°C
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Molecular Formula C7H5BrF2O
MDL Number MFCD00143258
CAS 202865-60-9
Health Hazard 1 Warning
Assay 99%
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.