missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2,5-difluoroanisole, 98%, ACROS Organics™

$43.31 - $43.31

Chemical Identifiers

CAS 202865-60-9
Molecular Formula C7H5BrF2O
Molecular Weight (g/mol) 223.02
MDL Number MFCD00143258
InChI Key OOTXNQBDWFJMNN-UHFFFAOYSA-N
Synonym 1-bromo-2,5-difluoro-4-methoxy-benzene, 4-bromo-2,5-difluoroanisole, acmc-1cm1k, benzene, 1-bromo-2,5-difluoro-4-methoxy, benzene,1-bromo-2,5-difluoro-4-methoxy, pubchem2653, timtec-bb sbb037912
PubChem CID 2724983
IUPAC Name 1-bromo-2,5-difluoro-4-methoxybenzene
SMILES COC1=CC(=C(C=C1F)Br)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC366490050
SDS View Documents
Thermo Fisher Scientific
366490050
5g Glass bottle
Each for $43.31
 
Description

Description

Specifications

Chemical Identifiers

202865-60-9
223.02
OOTXNQBDWFJMNN-UHFFFAOYSA-N
2724983
COC1=CC(=C(C=C1F)Br)F
C7H5BrF2O
MFCD00143258
1-bromo-2,5-difluoro-4-methoxy-benzene, 4-bromo-2,5-difluoroanisole, acmc-1cm1k, benzene, 1-bromo-2,5-difluoro-4-methoxy, benzene,1-bromo-2,5-difluoro-4-methoxy, pubchem2653, timtec-bb sbb037912
1-bromo-2,5-difluoro-4-methoxybenzene

Specifications

Authentic
White
5g
97.5
97.5% min. (GC)
MFCD00143258
OOTXNQBDWFJMNN-UHFFFAOYSA-N
1-bromo-2,5-difluoro-4-methoxybenzene
2724983
Crystals
4-Bromo-2, 5-difluoroanisole, 98%
Glass bottle
59.0°C to 63.0°C
202865-60-9
100.0
C7H5BrF2O
1-bromo-2,5-difluoro-4-methoxy-benzene, 4-bromo-2,5-difluoroanisole, acmc-1cm1k, benzene, 1-bromo-2,5-difluoro-4-methoxy, benzene,1-bromo-2,5-difluoro-4-methoxy, pubchem2653, timtec-bb sbb037912
COC1=CC(=C(C=C1F)Br)F
223.02
223.02
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS