missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2,5-difluoroanisole, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 366490050

 View more versions of this product

Catalog No. AC366490050



Description

Description

Specifications

Specifications

4-Bromo-2, 5-difluoroanisole
Glass bottle
59.0°C to 63.0°C
98%
97.5
97.5% min. (GC)
MFCD00143258
OOTXNQBDWFJMNN-UHFFFAOYSA-N
1-bromo-2,5-difluoro-4-methoxybenzene
2724983
Crystals
Authentic
White
5g
202865-60-9
100.0
C7H5BrF2O
1-bromo-2,5-difluoro-4-methoxy-benzene, 4-bromo-2,5-difluoroanisole, acmc-1cm1k, benzene, 1-bromo-2,5-difluoro-4-methoxy, benzene,1-bromo-2,5-difluoro-4-methoxy, pubchem2653, timtec-bb sbb037912
COC1=CC(=C(C=C1F)Br)F
223.02
223.02
98%

Chemical Identifiers

202865-60-9
223.02
OOTXNQBDWFJMNN-UHFFFAOYSA-N
2724983
COC1=CC(=C(C=C1F)Br)F
C7H5BrF2O
MFCD00143258
1-bromo-2,5-difluoro-4-methoxy-benzene, 4-bromo-2,5-difluoroanisole, acmc-1cm1k, benzene, 1-bromo-2,5-difluoro-4-methoxy, benzene,1-bromo-2,5-difluoro-4-methoxy, pubchem2653, timtec-bb sbb037912
1-bromo-2,5-difluoro-4-methoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents
Product Certifications