Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2,5-dichlorothiophene-3-sulfonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 4-Bromo-2,5-dichlorothiophene-3-sulfonyl chloride
CAS 166964-36-9
Molecular Formula C4BrCl3O2S2
Formula Weight 330.44
MDL Number MFCD00051664
UN Number UN3261
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1253303

  SDS
alfa aesar™
A1253303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1253314

  SDS
alfa aesar™
A1253314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 330.44
UN Number UN3261
CAS 166964-36-9
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Assay 97%
Chemical Name or Material 4-Bromo-2,5-dichlorothiophene-3-sulfonyl chloride
Molecular Formula C4BrCl3O2S2
MDL Number MFCD00051664
Boiling Point 119° to 121°C (1mmHg)
Health Hazard 1 Danger
Melting Point 67° to 71°C
Quantity 1g
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.