Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2,3-difluorobenzaldehyde, 98%

Chemical Identifiers

Chemical Name or Material 4-Bromo-2,3-difluorobenzaldehyde
CAS 644985-24-0
Formula Weight 221
MDL Number MFCD09258953
Beilstein 9551272
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6404403

 
alfa aesar™
H6404403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6404406

 
alfa aesar™
H6404406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64044MD

 
alfa aesar™
H64044MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221
Melting Point 66° to 67°C
Chemical Name or Material 4-Bromo-2,3-difluorobenzaldehyde
Physical Form Powder
Assay 98%
MDL Number MFCD09258953
CAS 644985-24-0
Beilstein 9551272
Color Off-white
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
4-Bromo-2,3-difluorobenzaldehyde is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.