Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2,3,5,6-tetrafluorobenzoyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 4-Bromo-2,3,5,6-tetrafluorobenzoyl chloride
CAS 122033-54-9
Molecular Formula C7BrClF4O
Formula Weight 291.43
MDL Number MFCD03001173
UN Number UN3265
Beilstein 7295811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2015303

  SDS
alfa aesar™
L2015303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2015306

  SDS
alfa aesar™
L2015306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2015314

  SDS
alfa aesar™
L2015314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 291.43
Chemical Name or Material 4-Bromo-2,3,5,6-tetrafluorobenzoyl chloride
Health Hazard 1 Danger
Assay 98%
Molecular Formula C7BrClF4O
Beilstein 7295811
MDL Number MFCD03001173
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Boiling Point 95° to 100°C
UN Number UN3265
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Quantity 1g
CAS 122033-54-9
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.