missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2,3,5,6-tetrafluorobenzoic acid, 97+%

$46.49 - $155.47

Chemical Identifiers

CAS 4707-24-8
Molecular Formula C7HBrF4O2
Formula Weight 272.98
MDL Number MFCD00075244
Chemical Name or Material 4-Bromo-2,3,5,6-tetrafluorobenzoic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3298203 SDS Alfa Aesar™
H3298203
1g Each for $46.49
AAH3298206 SDS Alfa Aesar™
H3298206
5g Each for $155.47
Specifications

Description

Applications
4-Bromo-2,3,5,6-tetrafluorobenzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
Description & Specifications

Chemical Identifiers

4707-24-8
272.98
4-Bromo-2,3,5,6-tetrafluorobenzoic acid
C7HBrF4O2
MFCD00075244

Specifications

4707-24-8
272.98
142° to 145°C
Odorless
≥97%
C7HBrF4O2
MFCD00075244
1g
4-Bromo-2,3,5,6-tetrafluorobenzoic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning