Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-(2-nitro-1-propenyl)thiophene, 97%

$81.81 - $287.80

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-(2-nitro-1-propenyl)thiophene
CAS 175205-19-3
Molecular Formula C7H6BrNO2S
Formula Weight 248.09
MDL Number MFCD00052971
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2506003

  SDS
alfa aesar™
B2506003
1g Each for $81.81

AAB2506006

  SDS
alfa aesar™
B2506006
5g Each for $287.80
Description & Specifications

Specifications

Formula Weight 248.09
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 175205-19-3
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Bromo-2-(2-nitro-1-propenyl)thiophene
Melting Point 88° to 90°C
Assay 97%
Molecular Formula C7H6BrNO2S
Health Hazard 1 Danger
MDL Number MFCD00052971
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.