missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-(1H-pyrazol-3-yl)phenol, TCI America™

$38.19 - $123.75

Chemical Identifiers

CAS 99067-15-9
Molecular Formula C9H7BrN2O
Molecular Weight (g/mol) 239.072
MDL Number MFCD03094026
InChI Key WAHULEHSSFNFQX-UHFFFAOYSA-N
Synonym 4-bromo-2-1h-pyrazol-3-yl phenol, 4-bromo-6-1,2-dihydropyrazol-3-ylidene cyclohexa-2,4-dien-1-one, 2-1h-pyrazole-3-yl-4-bromophenol, 4-bromo-2-2h-pyrazol-3-yl phenol
PubChem CID 6744501
IUPAC Name 4-bromo-6-(1,2-dihydropyrazol-3-ylidene)cyclohexa-2,4-dien-1-one
SMILES C1=CC(=O)C(=C2C=CNN2)C=C1Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B47431G View Documents TCI America
B47431G
1g Each for $38.19
 
B47435G View Documents TCI America
B47435G
5g Each for $123.75
 
Specifications

Chemical Identifiers

99067-15-9
239.072
WAHULEHSSFNFQX-UHFFFAOYSA-N
6744501
C1=CC(=O)C(=C2C=CNN2)C=C1Br
C9H7BrN2O
MFCD03094026
4-bromo-2-1h-pyrazol-3-yl phenol, 4-bromo-6-1,2-dihydropyrazol-3-ylidene cyclohexa-2,4-dien-1-one, 2-1h-pyrazole-3-yl-4-bromophenol, 4-bromo-2-2h-pyrazol-3-yl phenol
4-bromo-6-(1,2-dihydropyrazol-3-ylidene)cyclohexa-2,4-dien-1-one

Specifications

Purple-White
1g
C9H7BrN2O
4-bromo-2-1h-pyrazol-3-yl phenol, 4-bromo-6-1,2-dihydropyrazol-3-ylidene cyclohexa-2,4-dien-1-one, 2-1h-pyrazole-3-yl-4-bromophenol, 4-bromo-2-2h-pyrazol-3-yl phenol
C1=CC(=O)C(=C2C=CNN2)C=C1Br
239.072
239.07
≥98.0% (GC,T)
160°C
99067-15-9
MFCD03094026
WAHULEHSSFNFQX-UHFFFAOYSA-N
4-bromo-6-(1,2-dihydropyrazol-3-ylidene)cyclohexa-2,4-dien-1-one
6744501
Crystal-Powder at 20°C
4-Bromo-2-(1H-pyrazol-3-yl)phenol
Safety and Handling

Safety and Handling

TSCA : No