Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-(1H-pyrazol-3-yl)phenol, 98%

$34.95 - $454.08

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-(1H-pyrazol-3-yl)phenol
CAS 99067-15-9
Molecular Formula C9H7BrN2O
Formula Weight 239.07
MDL Number MFCD03094026
Synonym 3-(5-Bromo-2-hydroxyphenyl)-1H-pyrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2765303

  SDS
alfa aesar™
H2765303
1g Each for $34.95

AAH2765306

  SDS
alfa aesar™
H2765306
5g Each for $113.76

AAH2765314

  SDS
alfa aesar™
H2765314
25g Each for $454.08
Description & Specifications

Specifications

Formula Weight 239.07
Chemical Name or Material 4-Bromo-2-(1H-pyrazol-3-yl)phenol
Assay 98%
Molecular Formula C9H7BrN2O
MDL Number MFCD03094026
CAS 99067-15-9
Melting Point 159° to 160°C
Synonym 3-(5-Bromo-2-hydroxyphenyl)-1H-pyrazole
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.