Please login to your online account to display your discounted pricing

4-Bromo-1H-pyrazole-5-carbaldehyde, 95+%

Chemical Identifiers

Chemical Name or Material 4-Bromo-1H-pyrazole-5-carbaldehyde
Assay 95%
Assay Percent Range 95%
Molecular Formula C4H3BrN2O
Formula Weight 174.98g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RF03950CB

 
fisher scientific company
RF03950CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 250mg
Chemical Name or Material 4-Bromo-1H-pyrazole-5-carbaldehyde
CAS 287917-97-9
Assay 95%
Assay Percent Range 95%
Molecular Formula C4H3BrN2O
Formula Weight 174.98g/mol