Please login to your online account to display your discounted pricing

4-Bromo-1H-pyrazole-5-carbaldehyde, 95+%


Quantity 250mg

Manufacturer: fisher scientific company  RF03950CB

 View more versions of this product

Catalog No. RF03950CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 250mg
Chemical Name or Material 4-Bromo-1H-pyrazole-5-carbaldehyde
CAS 287917-97-9
Assay 95%
Assay Percent Range 95%
Molecular Formula C4H3BrN2O
Formula Weight 174.98g/mol

Specifications

Chemical Name or Material 4-Bromo-1H-pyrazole-5-carbaldehyde
Assay 95%
Assay Percent Range 95%
Molecular Formula C4H3BrN2O
Formula Weight 174.98g/mol