Please login to your online account to display your discounted pricing

4-bromo-1h-imidazole, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Bromo-1H-imidazole
Assay ≥96.0% (HPLC)
Assay Percent Range 97%
Molecular Formula C3H3BrN2
Formula Weight 146.97
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC376900010

  SDS
thermo fisher scientific
AC376900010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC376900050

  SDS
thermo fisher scientific
AC376900050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0% (HPLC)
Physical Form Crystalline Flakes or Powder
Formula Weight 146.97
Quantity 1g
Color Colorless to beige
Melting Point 126° to 132°C
Assay Percent Range 97%
Chemical Name or Material 4-Bromo-1H-imidazole
Molecular Formula C3H3BrN2
CAS 2302-25-2
Infrared Spectrum Authentic
Packaging Glass bottle