missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-1-naphthoic acid, 98%

$58.74 - $217.43

Chemical Identifiers

CAS 16650-55-8
Molecular Formula C11H7BrO2
Molecular Weight (g/mol) 251.079
MDL Number MFCD00094221
InChI Key FIJIPZQZVLCOMB-UHFFFAOYSA-N
Synonym 4-bromo-1-naphthoic acid, 4-bromo-1-naphthalenecarboxylic acid, 4-bromo-1-naphtalenecarboxylic acid, 1-naphthalenecarboxylic acid, 4-bromo, 4-bromo naphthoic acid, acmc-209dum, 4-bromo-naphthalic acid, 4-bromo-1-naphthoicacid, ksc175i3p, 1-naphthalenecarboxylicacid,4-bromo
PubChem CID 282591
IUPAC Name 4-bromonaphthalene-1-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CC=C2Br)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6482003 View Documents Alfa Aesar™
H6482003
1g Each for $58.74
 
AAH6482006 View Documents Alfa Aesar™
H6482006
5g Each for $217.43
 
Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

16650-55-8
251.079
FIJIPZQZVLCOMB-UHFFFAOYSA-N
282591
C1=CC=C2C(=C1)C(=CC=C2Br)C(=O)O
C11H7BrO2
MFCD00094221
4-bromo-1-naphthoic acid, 4-bromo-1-naphthalenecarboxylic acid, 4-bromo-1-naphtalenecarboxylic acid, 1-naphthalenecarboxylic acid, 4-bromo, 4-bromo naphthoic acid, acmc-209dum, 4-bromo-naphthalic acid, 4-bromo-1-naphthoicacid, ksc175i3p, 1-naphthalenecarboxylicacid,4-bromo
4-bromonaphthalene-1-carboxylic acid

Specifications

1g
C11H7BrO2
1950351
FIJIPZQZVLCOMB-UHFFFAOYSA-N
4-bromonaphthalene-1-carboxylic acid
282591
Solid
4-Bromo-1-naphthoic acid
16650-55-8
MFCD00094221
4-bromo-1-naphthoic acid, 4-bromo-1-naphthalenecarboxylic acid, 4-bromo-1-naphtalenecarboxylic acid, 1-naphthalenecarboxylic acid, 4-bromo, 4-bromo naphthoic acid, acmc-209dum, 4-bromo-naphthalic acid, 4-bromo-1-naphthoicacid, ksc175i3p, 1-naphthalenecarboxylicacid,4-bromo
C1=CC=C2C(=C1)C(=CC=C2Br)C(=O)O
251.079
251.08
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning