Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-1-methyl-1H-indole, 95%

Chemical Identifiers

Chemical Name or Material 4-Bromo-1-methyl-1H-indole
CAS 590417-55-3
Formula Weight 210.06
MDL Number MFCD08435875
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6653503

 
alfa aesar™
H6653503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66535MD

 
alfa aesar™
H66535MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.06
Chemical Name or Material 4-Bromo-1-methyl-1H-indole
Density 1.456g/mL
Color Colorless to yellow
Physical Form Liquid
Assay 95%
Quantity 1g
MDL Number MFCD08435875
CAS 590417-55-3
Refractive Index 1.6360