missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-1-methyl-1H-imidazole, 95%, ACROS Organics™

Chemical Identifiers

CAS 25676-75-9
Molecular Formula C4H5BrN2
Molecular Weight (g/mol) 161
InChI Key IOTSLMMLLXTNNH-UHFFFAOYSA-N
Synonym 1-methyl-4-bromo-1h-imidazole, 1h-imidazole, 4-bromo-1-methyl, 1h-imidazole,4-bromo-1-methyl, 4-bromanyl-1-methyl-imidazole, 4-bromo-1-methyl-1h-imidazole, 4-bromo-1-methyl-imidazole, acmc-1cjr4
PubChem CID 1277653
IUPAC Name 4-bromo-1-methylimidazole
SMILES CN1C=C(N=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC452430010
SDS View Documents
Thermo Fisher Scientific
AC452430010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC452430050
SDS View Documents
Thermo Fisher Scientific
AC452430050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

25676-75-9
161
1-methyl-4-bromo-1h-imidazole, 1h-imidazole, 4-bromo-1-methyl, 1h-imidazole,4-bromo-1-methyl, 4-bromanyl-1-methyl-imidazole, 4-bromo-1-methyl-1h-imidazole, 4-bromo-1-methyl-imidazole, acmc-1cjr4
4-bromo-1-methylimidazole
C4H5BrN2
IOTSLMMLLXTNNH-UHFFFAOYSA-N
1277653
CN1C=C(N=C1)Br

Specifications

>110°C
91°C to 93°C (0.1 hPa)
25676-75-9
C4H5BrN2
IOTSLMMLLXTNNH-UHFFFAOYSA-N
4-bromo-1-methylimidazole
1277653
95%
Glass Bottle
1g
94% min. (GC)
1-methyl-4-bromo-1h-imidazole, 1h-imidazole, 4-bromo-1-methyl, 1h-imidazole,4-bromo-1-methyl, 4-bromanyl-1-methyl-imidazole, 4-bromo-1-methyl-1h-imidazole, 4-bromo-1-methyl-imidazole, acmc-1cjr4
CN1C=C(N=C1)Br
161
161
4-Bromo-1-methyl-1H-imidazole
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

SDS