missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromo-1-methyl-1H-imidazole, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC452430050
Specifications
4-Bromo-1-methyl-1H-imidazole | |
Glass Bottle | |
5g | |
94% min. (GC) | |
1-methyl-4-bromo-1h-imidazole, 1h-imidazole, 4-bromo-1-methyl, 1h-imidazole,4-bromo-1-methyl, 4-bromanyl-1-methyl-imidazole, 4-bromo-1-methyl-1h-imidazole, 4-bromo-1-methyl-imidazole, acmc-1cjr4 | |
CN1C=C(N=C1)Br | |
161 | |
161 |
>110°C | |
91°C to 93°C (0.1 hPa) | |
25676-75-9 | |
C4H5BrN2 | |
IOTSLMMLLXTNNH-UHFFFAOYSA-N | |
4-bromo-1-methylimidazole | |
1277653 | |
95% |
Chemical Identifiers
25676-75-9 | |
161 | |
1-methyl-4-bromo-1h-imidazole, 1h-imidazole, 4-bromo-1-methyl, 1h-imidazole,4-bromo-1-methyl, 4-bromanyl-1-methyl-imidazole, 4-bromo-1-methyl-1h-imidazole, 4-bromo-1-methyl-imidazole, acmc-1cjr4 | |
4-bromo-1-methylimidazole |
C4H5BrN2 | |
IOTSLMMLLXTNNH-UHFFFAOYSA-N | |
1277653 | |
CN1C=C(N=C1)Br |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning