Please login to your online account to display your discounted pricing

4-Bromo-1-indanone, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC427700050

 View more versions of this product

Catalog No. AC427700050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 211.05
Quantity 5g
Color Orange-brown
Melting Point 94° to 98°C
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C9H7BrO
CAS 15115-60-3
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
NMR Consistent with structure
Assay Percent Range 97%
Chemical Name or Material 4-Bromo-1-indanone
MDL Number MFCD01719772
Packaging Glass bottle

Specifications

Chemical Name or Material 4-Bromo-1-indanone
CAS 15115-60-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H7BrO
Formula Weight 211.05
MDL Number MFCD01719772
Accessories

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents