missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-1-butyne, 97%

Manufacturer:  Alfa Aesar™ H5336706

 View more versions of this product

Catalog No. AAH5336706



Description

Description

Applications
4-Bromo-1-butyne is used for pharmaceutical intermediates.

Solubility
Immiscible with water.

Notes
Recommended storage in refrigerator. Keep away from sources of ignition. Incompatible with strong oxidizing agents.
Specifications

Specifications

4-Bromo-1-butyne
110°C
38771-21-0
HC≡CCH2CH2Br
UN1992
XLYOGWXIKVUXCL-UHFFFAOYSA-N
4-bromobut-1-yne
11073464
97%
24°C (75°F)
5g
C4H5Br
MFCD10000883
4-bromo-1-butyne, 1-bromo-3-butyne, 1-butyne, 4-bromo, 4-bromobutyne, 3-butynyl bromide, 1-bromo-3-butynol, 4-bromo-but-1-yne, acmc-1afup, 4-bromo-1-butyne, 1-butyne, 4-bromo-6ci,9ci
C#CCCBr
132.988
132.99

Chemical Identifiers

38771-21-0
132.988
XLYOGWXIKVUXCL-UHFFFAOYSA-N
11073464
C#CCCBr
C4H5Br
MFCD10000883
4-bromo-1-butyne, 1-bromo-3-butyne, 1-butyne, 4-bromo, 4-bromobutyne, 3-butynyl bromide, 1-bromo-3-butynol, 4-bromo-but-1-yne, acmc-1afup, 4-bromo-1-butyne, 1-butyne, 4-bromo-6ci,9ci
4-bromobut-1-yne
Safety and Handling

Safety and Handling

GHS H Statement
H301-H226-H317
Toxic if swallowed.
Flammable liquid and vapor.
May cause an allergic skin reaction.

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

Recommended Storage : 2° to 8°C

SDS Documents