missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-1-butanol, tech., cont. varying amounts of THF

Manufacturer:  Alfa Aesar™ 1680503

 View more versions of this product

Catalog No. AA1680503



Specifications

Specifications

4-Bromo-1-butanol
1.5035
63°C to 65°C (5mmHg)
1g
33036-62-3
Br(CH2)4OH
UN1987
SIJLYRDVTMMSIP-UHFFFAOYSA-N
4-bromobutan-1-ol
118388
Liquid
32° (91°F)
Moisture & Light Sensitive
Yellow
cont. varying amounts of THF
C4H9BrO
MFCD00015387
4-bromo-1-butanol, 1-butanol, 4-bromo, 4-bromobutanol, 4-bromo-butan-1-ol, 1-bromo-4-hydroxybutane, tetramethylene bromohydrin, 4-bromo-n-butanol, 1-bromobutan-4-ol, 4-bromo butan-1-ol, acmc-1axcv
C(CCBr)CO
153.019
153.02
Technical

Chemical Identifiers

33036-62-3
153.019
SIJLYRDVTMMSIP-UHFFFAOYSA-N
118388
C(CCBr)CO
C4H9BrO
MFCD00015387
4-bromo-1-butanol, 1-butanol, 4-bromo, 4-bromobutanol, 4-bromo-butan-1-ol, 1-bromo-4-hydroxybutane, tetramethylene bromohydrin, 4-bromo-n-butanol, 1-bromobutan-4-ol, 4-bromo butan-1-ol, acmc-1axcv
4-bromobutan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H226-H351-H315-H319
Flammable liquid and vapor.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

storageNote1 : −30°C to −10°C

SDS Documents