missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-1,3,5-trimethylpyrazole, TCI America™

$48.89 - $140.56

Chemical Identifiers

CAS 15801-69-1
Molecular Formula C6H9BrN2
Molecular Weight (g/mol) 189.056
MDL Number MFCD00052532
InChI Key UNTQXOJGXGRHMG-UHFFFAOYSA-N
Synonym 4-bromo-1,3,5-trimethyl-1h-pyrazole, 4-bromo-2,3,5-trimethylpyrazole, 4-bromo-1,3,5-trimethyl-2h-pyrazole, pubchem20507, acmc-209dhc, buttpark 34\09-42, 4-bromo-1,3,5-trimethylpyrazole, 4-bromo-1,3,5-trimethyl-pyrazole, 4-bromo-1,3,5-trimethylpyrazole, 4-bromo-1,3,5-trimethyl pyrazole
PubChem CID 519181
IUPAC Name 4-bromo-1,3,5-trimethylpyrazole
SMILES CC1=C(C(=NN1C)C)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B47441G View Documents TCI America
B47441G
1g Each for $48.89
 
B47445G View Documents TCI America
B47445G
5g Each for $140.56
 
Specifications

Chemical Identifiers

15801-69-1
189.056
UNTQXOJGXGRHMG-UHFFFAOYSA-N
519181
CC1=C(C(=NN1C)C)Br
C6H9BrN2
MFCD00052532
4-bromo-1,3,5-trimethyl-1h-pyrazole, 4-bromo-2,3,5-trimethylpyrazole, 4-bromo-1,3,5-trimethyl-2h-pyrazole, pubchem20507, acmc-209dhc, buttpark 34\09-42, 4-bromo-1,3,5-trimethylpyrazole, 4-bromo-1,3,5-trimethyl-pyrazole, 4-bromo-1,3,5-trimethylpyrazole, 4-bromo-1,3,5-trimethyl pyrazole
4-bromo-1,3,5-trimethylpyrazole

Specifications

97°C
36°C
15801-69-1
MFCD00052532
UNTQXOJGXGRHMG-UHFFFAOYSA-N
4-bromo-1,3,5-trimethylpyrazole
519181
Crystal-Lump at 20°C
4-Bromo-1,3,5-trimethylpyrazole
White-Yellow
1g
C6H9BrN2
4-bromo-1,3,5-trimethyl-1h-pyrazole, 4-bromo-2,3,5-trimethylpyrazole, 4-bromo-1,3,5-trimethyl-2h-pyrazole, pubchem20507, acmc-209dhc, buttpark 34\09-42, 4-bromo-1,3,5-trimethylpyrazole, 4-bromo-1,3,5-trimethyl-pyrazole, 4-bromo-1,3,5-trimethylpyrazole, 4-bromo-1,3,5-trimethyl pyrazole
CC1=C(C(=NN1C)C)Br
189.056
189.06
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No